Volumetric flasks, class A, amber markings, BLAUBRAND® ETERNA
Leverandør: Brand
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2.36853 2.36845 2.36851 2.36843 2.36838 2.36848 2.36850 2.36854 2.36842 2.36852 2.36849 2.36847 2.36846
BRND2.36853EA 638 DKK
BRND2.36853 BRND2.36845 BRND2.36851 BRND2.36843 BRND2.36838 BRND2.36848 BRND2.36850 BRND2.36854 BRND2.36842 BRND2.36852 BRND2.36849 BRND2.36847 BRND2.36846
Volumetric flasks, class A, amber markings, BLAUBRAND® ETERNA
Kolber Volumetriske kolber
Borosilicate glass 3.3, with PP stopper.
  • Calibrated to contain (TC, In)
  • Mark and inscriptions in ETERNA amber stain
  • DE-M mærkning
  • Total order from one batch, supplied with batch certificate
*Wide neck
Certificeringer: DIN EN ISO 1042
Bestillingsoplysninger: Delivered with batch number and enclosed one batch certificate per packing unit of the manufacturer. On demand also available with individual certificate or DAkkS calibration certificate.