Gown racks for full length overalls
Leverandør: Palbam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
GR08CMP GR16CMP GR24CMP GR32CMP GR40CMP GR24CMP-C GR08CMP-C GR16CMP-C GR16CMP-1750MM
134-0490EA 17640 DKK
134-0490 134-0491 134-0492 134-0493 134-0494 PALBGR24CMP-C PALBGR08CMP-C PALBGR16CMP-C PALBGR16CMP-1750MM
Gown racks for full length overalls
Møbler Tøjstativer
Gown racks in stainless steel 304L with electropolished finish
  • Rack design holds each hanger at exactly the same angle and spacing ensuring a uniform appearance, creating an image of orderliness and allowing sufficient laminar flow
  • Optional mirrors (purchased separately) can be connected to side panels, enhancing both aesthetics and functionality
  • Each gowning rack is supplied with hangers, labelling numbers and leveller legs with integral covers
  • Hangers - the thickness of the material ensures non deforming
Leveringsinformation: Typically availableMore Product Information