Cartridge filters, Fluorogard® AT
Leverandør: ENTEGRIS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
CTFG0STPE CTFV01TPEPW CTFG01TPEKPW CTFV0STPE CTFZ0STPE CTFV01TK1
ENTECTFG0STPEEA 0 DKK
ENTECTFG0STPE ENTECTFV01TPEPW ENTECTFG01TPEKPW ENTECTFV0STPE ENTECTFZ0STPE ENTECTFV01TK1
Cartridge filters, Fluorogard® AT
Filtre Filterpatroner
These filters are designed to consistantly remove particles from a whide range of acids, bases and other process materials. Ideal for medium to high flow applications, they deliver a low cost of ownership (COO) as a dry device or prewet product ready for quick installation. Prewet cartridges prevent alcohol-chemical interactions, avoid potential contamination sources and minimise process downtime.
  • Ideal for higher temperature applications
  • Broad range of chemical compatibility
  • Manufactured in ISO 9001 certified facilities under cleanroom conditions and documentation control
Housing: PTFE, membrane: pleated hydrophobic PTFE
Maximum operating temperature: 150 °C
Maximum forward differential pressure: 5,1 bar at 25 °C; 0,51 bar at 150 °C
Maximum reverse differential pressure: 3,5 bar at 25 °C