Microtitre plates, 96 well, PS
Leverandør: SEMADENI
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
4510 4513
SEMA4510EA 2370 DKK
SEMA4510 SEMA4513
Microtitre plates, 96 well, PS
Mikroplader HTS mikroplader
PS, optically clear. Suitable for use with multi-channel pipettes. Alphanumeric labelling.