Culture tubes, disposable, KIMBLE®
Leverandør: DWK Life Sciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SP73500-25150 SP73500-18150 SP73500-16100A
392-0368EA 1910 DKK
392-0368 392-0367 SCERSP73500-16100A
Culture tubes, disposable, KIMBLE®
Rør Dyrkningsrør
Culture tubes made from 51 expansion borosilicate glass that offer excellent chemical resistance against sodium leaching, a factor common in soda lime glass products.
  • Features sturdy, uniform bottoms and consistent lengths
  • Suitable for cell washing procedures
  • Plain and disposable
Certificeringer: Glass meets ASTM E438, Type I, Class B requirements. Designed from ASTM E890 specifications.
Emballering: Tubes are packed in convenient trays that are shrink-wrapped in plastic film, with tubes remaining in constant alignment for protection against in-More Product Information