Cold resistant gloves, ActivArmr® 78-102 / 78-202
Leverandør: Ansell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
78-103/9 78-202/7 78-202/9 78-203/7 78-203/9 78-102/7 78-102/9 78-103/7
112-4967EA 465 DKK
112-4967 112-4996 112-4997 112-4998 112-4999 112-4964 112-4965 112-4966
Cold resistant gloves, ActivArmr® 78-102 / 78-202
Handsker
These knitted acrylic/spandex gloves offer very good thermal insulation. They are available with or without PVC dot coating on the palm.
  • Models with micro dots on the palm are excellent for applications where additional grip is required
  • Can be used as undergloves with liquid protective or cut protective gloves
  • Latex-free: No risk of Type 1 allergies
Ideal for a very wide range of food processing applications (storage areas, handling of frozen products, transportation).
Gauge: 10
Colour: Blue
78-102/78-103: EN 388 (113X), EN 511 (010), Cat. II 78-202/78-203: EN 388 (313X), EN 511 (010), Cat. II Food contact approved
Certificeringer: 78-102/78-103: EN 388:2016 (113XB), EN 511 (010), Cat. II
78-202/78-203: EN 388:2016 (313XB), EN 511 (010), Cat. II
Food contact approved
Emballering: 78-102/78-202: 1 pair in a bag.
78-103/78-203: 12 pairs in a bag.