Inverted microscopes, AE31E
Leverandør: MOTIC
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1100100200565 1100100200576 1100100200594 P31L P31HD P31LED P31S P31FLUOR
630-2518EA 30870 DKK
630-2518 630-2519 630-2521 MOTIP31L 630-2971 MOTIP31LED MOTIP31S MOTIP31FLUOR
Inverted microscopes, AE31E
Mikroskoper
The AE31E has been designed with an extended focus for practical aspects.
  • Automatic on/off setting via infrared sensor
  • Light memory recalls the last intensity, when changing objective position
  • Halogen interchangeable with an LED module to be purchased separately (30 W versions)
NOTE: Options such as fluorescence are available − please contact your local Avantor sales office.
Leveringsinformation: Phase objectives supplied as standard (4×, Ph 10×, LWD Ph 20×) work all three with the same Ph1 ring.
Bestil nu
Specifikationer
Environmentally Preferable
Head Siedentopf, 45° vinklet Siedentopf, 45° vinklet
Eyepieces WF 10×/22 mm med diopterjustering og øjekop af gummi WF 10×/22 mm med diopterjustering og øjekop af gummi
Nosepiece 5 positioner, inclined 5 positioner, inclined
Objectives 4×, ph10×, phLWD 20×, ph LWD 40× 4×, ph10×, phLWD 20×, ph LWD 40×
Stage Hard coated with metal and glass stage inserts
Condenser N.A 0,30 with extra-long working distance (72 mm) and diaphragm N.A 0,30 with extra-long working distance (72 mm) and diaphragm
Fokuseringssystem Koaksial grov og finindstilling Koaksial grov og finindstilling
Illumination Halogen (Köhler) Halogen (Köhler)