Powder Sampler, SteriWare®
Leverandør: Sampling Systems
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
8240H-1000 8240H-1000S
SAMP8240H-1000EA 4620 DKK
SAMP8240H-1000 SAMP8240H-1000S
Powder Sampler, SteriWare®
Prøvetagere Prøvetagere til engangsbrug
Designed to collect single large volume sample from a single known point of free flowing powders from drums, kegs, and most containers.
  • Single use with gamma irradiation
  • Assembled and packed in a cleanroom environment - Class 100000
  • BSE/TSE free as well as full batch traceability
Certificeringer: FDA and EU 10/2011 conformity, ATEX rated.