Gas washing bottles, with head, BOLA
Leverandør: Bohlender
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
N1662-44 N1662-14 N1662-24 N1662-34
BOHLN1662-44EA 2280 DKK
BOHLN1662-44 BOHLN1662-14 BOHLN1662-24 BOHLN1662-34
Gas washing bottles, with head, BOLA
Flasker Gasvaskeflasker
Clear borosilicate glass 3.3 bottle with FEP inlet tube.
  • Chemical resistant
  • Temperature resistant from −200 to +250 °C
Glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.