Cleanroom tables, BLAUTOUCH
Leverandør: LABORIAL
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
AZ1AAABABE00AB AZ1AAABABEA0AA AZ1AAABABEA0AB AZ1AAABABE00AA BZ1AAAEABE00BC BZ1AAAEABEA0BC
132-1087EA 0 DKK
132-1087 132-0983 132-1088 132-0982 132-1089 132-1090
Cleanroom tables, BLAUTOUCH
Møbler Borde
Innovative interactive worktop for laboratories, cleanrooms and healthcare. A hygienic design based on GMP guidlines which consists of a continuous, tempered glass surface on phenolic resin laminate in which is embedded a touch control screen, to provide an interactive worktop, all supported on a metallic structure. At the same time, it works as a regular worktop and solves the disinfection and contamination problems associated with the use of tablets, notebooks, PCs, keyboards and mouse in cleanrooms and other controlled environments. It allows access to any laboratory software such as electronic notebooks, LIMS, Wikis, analysis software, Office and others, on the laboratory workbench, whilst maintaining an aseptic environment. The interactive worktop can help to reduce traffic, in and out of critical areas in which the use of handheld computer devices are restricted or prohibited.
  • Easy to clean and decontaminate
  • Allows the use of gloves
  • Avoids biological, chemical and radioactive contamination
  • High chemical and mechanical shock resistance
  • High levels of safety and structural stability
  • Optional integrated 2D barcode scanner for facilitating good sample data identification
  • Saves space and provides a tool towards achieving the ideal 'paperless' laboratory
  • Allows laboratories to comply with safety guidelines
  • Modular - can integrate into similar modular Laborial cleanroom furniture
Material:
Worktop surface in tempered glass, on top of a second layer of phenolic resins
Sealed electronic components enclosure in ST12 steel
'C' frame made in ST37-2 steel
Linking frame in ST12 cold rolled steel
Powder coating with high chemical resistance based on a mixture of epoxy polyester resins
All metallic components with powder coating
2 M10 levelers in steel and polyethylene stoppers, at the base
Touch specifications:
Touch technology: State of the art, projected capacitance technology
Touch accuracy: 1 mm relative
Touch latency: 22 ms
Input method: Finger or gloved finger
Touch gestures: Microsoft Windows 7 and Windows 8 touch gestures – tap and double-tap, selection/drag, press and tap with second finger, zoom, rotate, press and hold, flicks
Display specifications:
Display technology: LCD-LED
Display size: 27"
Native resolution: Full-HD 1920×1080
Display colours: 16,7 million
Brightness: 250 cd/m²
Contrast ratio: 5000000:1
Viewing angle: 178°/168°
Embedded computer specifications:
CPU: Intel™ core i3 330M @ 1,20 GHz or equivalent
RAM: 2 GB RAM DDR3 1066 MHz
Hard disk: 320/500 GB SATA
Wi-Fi: Compatible WPA/802.11 a/b/g/n - IEEE 802.1x
Operating system: Windows 7 32/64-bit and Windows 8 32/64-bit (optional)
I/O connections: Protected ports for 3×USB A and 1×RJ45
Environment specifications:
Operating temperature: 0 to +25 °C
Relative humidity: 20 to 80% (non condensing)
Certificeringer: Workbench developed according to EN 13150
Developed according to GMP guidelines 'EudraLex Vol 4, Chapter 3: Premises and Equipment'
ST37-2 steel according to DIN 17100
ST12 steel according to DIN 1623
Powder coating according to the finishing’s European Standards, GBS, Qualicoat Class 1 and BS6496:1984, BS6497:1984.