Erlenmeyer flasks, narrow neck, with screw cap, Corning®
Leverandør: Corning
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
4985P-1L 4985P-500 4985P-250
CORN4985P-1LEA 431 DKK
CORN4985P-1L CORN4985P-500 CORN4985P-250
Erlenmeyer flasks, narrow neck, with screw cap, Corning®
Kolber Erlenmeyer-kolber
PP, translucent.
  • Very good chemical resistance
  • Very good temperature resistance
With blue, easy to read graduations. Autoclavable. The flasks feature a stopper bed and can also be sealed with standard ground joint polypropylene stoppers.