Sharps Container
Leverandør: USON PLAST
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
720 715
USON720EA 5110 DKK
USON720 USON715
Sharps Container
Beholdere Sharps containere
Container, made of PP, yellow.
  • Made of environmentally friendly, strong and durable Polypropylene
  • Design and manufacturing secure a high degree of safety
  • Reversible closure can be safely fixed
  • Containers are waterproof when closed
These sharp containers is designed for hospitals and health care institutions to dispose of syringes, scalpels and other types of sharps. Designed to maximise safety whilst still maintaining a barrier to protect the user and prevent spillage during use.
Different lid designs give the possibility to discharge needles, syringes and other types of sharps.
Certificeringer: Meet the UN requirement no. 3291 regarding packaging approved for transportation of dangerous goods (waste/disposal).