HelixMark® Tubing, Silicone, Platinum Cured, ELASTOSIL
Leverandør: Freudenberg Medical Europe GmbH
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
60-831-01 60-831-02 60-831-03 60-831-04 60-831-06 60-831-07 60-831-05
228-0851EA 0 DKK
228-0851 228-0852 228-0853 228-1162 228-1163 228-1164 228-0854
HelixMark® Tubing, Silicone, Platinum Cured, ELASTOSIL
Slanger Overførselsslange
HelixMark® standard silicone tubing meets the needs of a broad spectrum of medical and laboratory applications and are universally applicable for the transfer of fluids under standard requirements. The tubing is individually packed in 15 m coils.
  • Manufactured from platinum-cured silicone
  • Typical hardness: 80 Shore
  • Production and packaging in ISO Class 8 cleanrooms
  • Sterilisable in autoclaves, with ethylene oxide and by gamma irradiation
  • Seamless batch traceability
  • Double-packed
All HelixMark® products are manufactured in cleanrooms in compliance with the requirements of the medtech and pharmaceutical industry. The following criteria apply to all articles of this programme:
Materials approved in accordance with USP Class VI, ISO 10993-1, FDA 21CFR 177.2600 and EP 3.1.9
Utilisation of platinum-cured silicone
Production and packaging in ISO Class 8 cleanrooms
Certified manufacturing in compliance with ISO 13485 quality standards
Sterilisable in autoclaves, with ethylene oxide and by gamma irradiation
Supplied with material certificate in every packaging unit
Seamless batch traceability
Double-packed
Off-the-shelf availability to guarantee short delivery times
Customer-specific solutions available outside the HelixMark® standard product portfolio
Certificeringer: ISO 10993-1. FDA 21CFR 177-2600. USP Class VI. EP 3.1.9.