High temperature cleanroom tapes
Leverandør: ULTRATAPE
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
0156BZ038-P3S 0156BZ075-P3SU
UTAP0156BZ038-P3SEA 257 DKK
UTAP0156BZ038-P3S UTAP0156BZ075-P3SU
High temperature cleanroom tapes
Tape
These high temperature cleanroom tapes are made of polyimide with silicone adhesive.
  • Removes without adhesive residue
  • Resistant to tearing and punctures
  • For use in cleanroom
Length: 32,9 m
Colour: Bronze