Isolator gloves, AlphaTec® Neoprene
Leverandør: Ansell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
55-305/10 55-301/10 55-301/9 55-300/10 55-300/8 55-300/9 55-302/10 55-303/10 55-306/10 55-307/10 55-308/10
112-3468EA 1630 DKK
112-3468 112-3460 112-3461 112-3457 112-3458 112-3459 112-3462 112-3464 112-3470 112-3472 112-3474
Isolator gloves, AlphaTec® Neoprene
Handsker
These ambidextrous gauntlets are made ​​of neoprene without an inner lining. They provide protection against chemical and mechanical hazards as well as against biohazards. For use with most available cleanroom glove boxes.
  • Highly resistant to most acids, oils, alcohols, lubricants and hydrocarbons
  • Beaded cuffs for easier donning
  • Good abrasion resistance
  • Liquid-tight
Ideal for chemical and medical research, electronics, pharmaceuticals, sandblasting, and steam cleaning.
Colour: Black
EN 374 (ACJKL); Cat. III; AQL 0,65 Thickness 0,33/0,51 mm: EN 388 (2100) Thickness 0,76 mm: EN 388 (3101)
Certificeringer: EN ISO 374-1 Type A (JKLOPT), EN ISO 374-5 VIRUS; Cat. III; AQL 0,65
Thickness 0,33/0,51 mm: EN 388:2016 (2100A)
Thickness 0,76 mm: EN 388:2016 (3101A).