Isolator gloves, AlphaTec® latex
Leverandør: Ansell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
55-107/10 55-104/10 55-109/10 55-100/10 55-100/8 55-100/9 55-101/10 55-101/8 55-101/9 55-105/10 55-110/10
112-3447EA 1330 DKK
112-3447 112-3441 112-3451 112-3431 112-3432 112-3433 112-3434 112-3435 112-3436 112-3443 112-3452
Isolator gloves, AlphaTec® latex
Handsker
These ambidextrous gauntlets are made ​​of natural rubber latex without an inner lining. They provide protection against chemical and mechanical hazards as well as against biohazards. For use with most available cleanroom glove boxes.
  • Highly resistant to most acids, alkalis, alcohols, aldehydes and ketones
  • Good tear resistance
  • Beaded cuffs for easier donning
  • Liquid-tight
Ideal for chemical and medical research, electronics, pharmaceuticals, sandblasting, and steam cleaning.
Colour: Natural
EN 374; Cat. III; AQL 0,65
Thickness 0,51 mm: EN 388 (3010)
Thickness 0,76 mm: EN 388 (3121)
Certificeringer: EN ISO 374-1 Type A (KLNOPT), EN ISO 374-5 VIRUS; Cat. III; AQL 0,65
Thickness 0,51 mm: EN 388:2016 (2020X)
Thickness 0,76 mm: EN 388:2016 (2121A)