Marker pens, sterile, BioClean Permaflow™
Leverandør: Nitritex
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
S-BPFP-BL1 S-BPFP-RD1 S-BPFP-BK1
NITRS-BPFP-BL1EA 348 DKK
NITRS-BPFP-BL1 NITRS-BPFP-RD1 NITRS-BPFP-BK1
Marker pens, sterile, BioClean Permaflow™
Penne Permanente markører
BioClean Permaflow™ Pens are processed to ensure ISO Class 4 and EU GMP Grade A compatibility and autoseal ink prevents drying.
  • Fade and water resistant
  • Quick drying
  • Non-toxic
Emballering: Ten pieces double bagged