VWR® Adapt-A Rack®, Test Tube Racks
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
525-0702EA 153 DKK
525-0702 525-0703 525-0704 525-0705 525-0706 525-0707 525-0708 525-0709
VWR® Adapt-A Rack®, Test Tube Racks
Stativer Tube Racks
POM.
  • Capable of holding tubes from 12 to 30 mm (approx. 5 to 50 ml) in the same well
  • The slot can hold round, flat, conical and even skirted tubes
  • Link together to create as large or as small a rack as needed
  • A large range of colours available for end user, application or department identification
  • Open sides allow easy label viewing
An innovative flexible multi-tube rack that is capable of holding different sized tubes in each available slot, as well as literally adapting to your tube size to hold firmly in the upright position. Each rack has four available slots which can accommodate tubes from 12 to 30 mm in Ø. Racks in a single colour or two tone colours are available.
Leveringsinformation: Each rack supplied in a pack of 2.