Foliodress® Suit Surgical Suits
Leverandør: HARTMANN PAUL
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
9925214 9925194
HART9925214EA 2230 DKK
HART9925214 HART9925194
Foliodress® Suit Surgical Suits
Engangsbeklædning
This disposable surgical clothing consist of one pair of trousers and a tunic made ​​of breathable, non woven fabric.
  • Tunic with V-neck, short sleeves and pocket
  • Trousers with straight legs and ties at the waist
  • Liquid repellent, high resistance to moisture
  • Antibacterial
  • Abrasion resistant and low linting
EN 13795 (parts 1 -3)
Certificeringer: EN 13795 (parts 1 - 3).