Frame and corrective lens for goggles
Leverandør: UNIVET
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
3PA299 042.120M 601.OP.00.99 042.060 5X1.K1.00.00 5X1.K2.03.00 6X1O.00.99 6X3F.01.00
UNVT3PA299EA 166 DKK
UNVT3PA299 UNVT042.120M UNVT601.OP.00.99 UNVT042.060 UNVT5X1.K1.00.00 UNVT5X1.K2.03.00 UNVT6X1O.00.99 UNVT6X3F.01.00
Frame and corrective lens for goggles
Beskyttelsesbriller
The RX insert is a special support that allows to equip normal eyewear with corrective lenses.
  • Device permits to merge into a single product increased protection to the correction of visual defects
  • Insert is strictly personal as it is built upon specific request in accordance with medical prescription
  • Can be easily applied and removed from the protective device