SPE tubes, Supelclean™ ENVI-Carb™, Supelco®
Leverandør: Merck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
57088 57092 57094 57109-U 57127-U 57128 57130
57088.EA 1420 DKK
57088. 57092. 57094. 57109-U 57127-U 57128. 57130.
SPE tubes, Supelclean™ ENVI-Carb™, Supelco®
Kromatografikolonner
SPE tubes made from graphitised non-porous carbon.
  • Extreme affinity for organic polar and non-polar compounds from both non-polar and polar matrices when used under reversed-phase conditions
  • Carbon surface comprised of hexagonal ring structures, interconnected and layered into graphitic sheets
  • Non-porous nature of the carbon phase allows for rapid processing, adsorption does not require analyte dispersion into solid phase pores
  • Independent investigators have found ENVI-Carb™ extremely useful for the rapid sample preparation of over 200 pesticides from various matrices including ground water, fruits and vegetables