Disinfectants, ClearKlens DE
Leverandør: Diversey
Ratings:
Gennemsntilig vurdering: 3.0 (Bedømmelse)
Samlet vurdering
Gennemsntilig vurdering
Var denne bedømmelse brugbar? Ja Nej
Ja: 0 Nej: 0 
Ja, jeg anbefaler
Bonjour, j'ai besoin d'obtenir la Fiche de Données de Sécurité du produit ClearKlens. Je vous remercie de me transférer ce document. Bonne journée. Delphine PETIT
Svar
vwr.com medarbejder Offentliggjort 29/05/2018
Vous recevrez la Fiche de Données de Sécurité du produit ClearKlens. par email.
6069643 100862174
141-1217EA 1070 DKK
141-1217 DIVY100862174
Disinfectants, ClearKlens DE
Renseopløsninger Disinfektanter
ClearKlens DE is a ready to use sterile disinfectant with 70% ethanol and 30% WFI (water for injection). It has the broad spectrum of activity against both gram-positive and gram-negative bacteria, yeasts and fungi. Can be used for the disinfection of work surfaces and process equipment in cleanrooms. Designed for spraying and wiping applications.
  • 0,2 μm filtered, gamma irradiated (900 ml) or sterile filled (5 L pack size) and double-bagged
  • Detex-labelled for proof of irradiation (900 ml)
  • Supplied with Certificates of Analysis, irradiation, endotoxin and sterility where applicable
  • 900 ml trigger spray pack incorporates anti-suck technology
Certificeringer: Bactericide EN 1276; Fungicide EN 1650; Bactericide and Fungicide EN 13697.
Advarsel: Use biocides safely. Always read the label and product information before use.