Sterile chemotherapy prep bench mat, BioClean ChemPrep™
Leverandør: Nitritex
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
S-BCPM-4025B S-BCPM-5640B
115-1880EA 7190 DKK
115-1880 115-1938
Sterile chemotherapy prep bench mat, BioClean ChemPrep™
Bordpapir Bordpapir
The sterile chemotherapy prep mat from BioClean is highly absorbent.
  • Three-layer construction
  • Mesh upper layer for even distribution, highly absorbent middle layer, impermeable bottom layer
  • Sterilised by gamma radiation, low-linting, latex-free
Certificeringer: ISO 9001:2008, EU GMP Grade A compatible
Advarsel: To be stored in a dry, cool place (up to 40 °C) away from direct sunlight and fluorescent light.