Anti-slip multipurpose reels, Original
Leverandør: DYCEM
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
NS03/L1/1 NS03/L1/2 NS03/L1/3 NS03/L1/5 NS03/L1/6 NS03/L2/1 NS03/L2/2 NS03/L2/3 NS03/L2/5 NS03/L2/8 NS03/L2/C NS03/L9/1 NS03/L9/2 NS03/L9/3 NS03/L9/5 NS03/L9/8 NS03/S1/3 NS03/S1/8 NS03/S1/C NS03/S2/1 NS03/S2/2 NS03/S2/3 NS03/S2/5 NS03/S2/8 NS03/S9/1 NS03/S9/2 NS03/S9/3 NS03/S9/5 NS03/S9/8 NS03/S9/C NS03/S1/1 NS03/S1/2 NS03/L1/8 NS03/S1/5 NS03/S9/6
DYCENS03/L1/1EA 405 DKK
DYCENS03/L1/1 DYCENS03/L1/2 DYCENS03/L1/3 DYCENS03/L1/5 DYCENS03/L1/6 DYCENS03/L2/1 DYCENS03/L2/2 DYCENS03/L2/3 DYCENS03/L2/5 DYCENS03/L2/8 DYCENS03/L2/C DYCENS03/L9/1 DYCENS03/L9/2 DYCENS03/L9/3 DYCENS03/L9/5 DYCENS03/L9/8 DYCENS03/S1/3 DYCENS03/S1/8 DYCENS03/S1/C DYCENS03/S2/1 DYCENS03/S2/2 DYCENS03/S2/3 DYCENS03/S2/5 DYCENS03/S2/8 DYCENS03/S9/1 DYCENS03/S9/2 DYCENS03/S9/3 DYCENS03/S9/5 DYCENS03/S9/8 DYCENS03/S9/C DYCENS03/S1/1 DYCENS03/S1/2 DYCENS03/L1/8 DYCENS03/S1/5 DYCENS03/S9/6
Anti-slip multipurpose reels, Original
Bordpapir Bordpapir
Multi-purpose reels made from durable polymeric material coated with non-slip properties. Can be cut into sections or shapes and it is proven to grip objects up to a 45° angle.
  • Antimicrobial
  • Long lasting, efficient and discreet
  • Non-toxic, latex free
Available in different sizes, colours and thickness.