Washable cleanroom floor mats, WorkZone
Leverandør: DYCEM
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
CZ02/12/CH CZ02/12/GN CZ02/20/CH CZ02/20/GN
DYCECZ02/12/CHEA 15320 DKK
DYCECZ02/12/CH DYCECZ02/12/GN DYCECZ02/20/CH DYCECZ02/20/GN
Washable cleanroom floor mats, WorkZone
Måtter Permanente Gulvmåtter
Specially designed mat to remove contamination from areas subject to heavy wheeled traffic. It effectively attracts, collects and retains contamination from the wheels of forklifts, pallet trucks, large trolleys and carts, preventing particles and microbes from getting into critical areas.
  • Flexible to be installed at different sized and shaped areas
  • Prevents 99% of contamination
  • Available as flame retardant or coving and inlaid
  • Also available in two different edging options: heavy duty aluminium edging or a lower profile stainless steel
Colour: Dark grey or white