ESD sikkerhedssko, med snørebånd, Uvex1, 8544
Leverandør: UVEX
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
8544.8/42 8544.8/44 8544.8/35 8544.8/36 8544.8/37 8544.8/38 8544.8/39 8544.8/40 8544.8/41 8544.8/45 8544.8/43 8544.8/46 8544.8/47 8544.8/48 8544.8/49 8544.8/50 8544.8/51 8544.8/52 8544.7/44 8544.7/42 8544.7/49 8544.7/50 8544.7/51 8544.7/52 8544.7/43 8544.7/39 8544.7/35 8544.7/36 8544.7/37 8544.7/38 8544.7/46 8544.7/47 8544.7/48 8544.7/41 8544.7/45 8544.7/40 8544.9/40 8544.9/41 8544.9/42 8544.9/44 8544.9/45 8544.9/46 8544.9/47 8544.9/48 8544.9/49 8544.9/50 8544.9/51 8544.9/52 8544.9/37 8544.9/35 8544.9/36 8544.9/38 8544.9/43 8544.9/39 8544.0/35 8544.0/36 8544.0/37 8544.0/38 8544.0/39 8544.0/40 8544.0/41 8544.0/42 8544.0/43 8544.0/44 8544.0/45 8544.0/46 8544.0/47 8544.0/48 8544.0/49 8544.0/50 8544.0/51 8544.0/52
114-5557EA 974 DKK
114-5557 114-5559 114-5550 114-5551 114-5552 114-5553 114-5554 114-5555 114-5556 114-5560 114-5558 114-5561 114-5562 114-5563 114-5564 114-5565 114-5566 114-5567 114-6200 114-6198 114-6205 114-6206 114-6207 114-6208 114-6199 114-6195 114-6191 114-6192 114-6193 114-6194 114-6202 114-6203 114-6204 114-6197 114-6201 114-6196 114-7866 114-7979 114-7980 114-7982 114-7983 114-7984 114-7985 114-7986 114-7987 114-7988 114-7989 114-7990 114-7976 114-7974 114-7975 114-7977 114-7981 114-7978 UVEX8544.0/35 UVEX8544.0/36 UVEX8544.0/37 UVEX8544.0/38 UVEX8544.0/39 UVEX8544.0/40 UVEX8544.0/41 UVEX8544.0/42 UVEX8544.0/43 UVEX8544.0/44 UVEX8544.0/45 UVEX8544.0/46 UVEX8544.0/47 UVEX8544.0/48 UVEX8544.0/49 UVEX8544.0/50 UVEX8544.0/51 UVEX8544.0/52
ESD sikkerhedssko, med snørebånd, Uvex1, 8544
Sko og støvler
Metal-free, extremely lightweight, flexible and multifunctional safety shoe made from breathable water resistant high-tech micro velour upper, soft padded tongue and collar, and a dual-density PUR sole (oil and petrol resistant, anti-static) free of lacquer wetting inhibitors such as silicones or phthalates.
  • Climazone technology for improved comfort
  • Breathable distance mesh lining
  • Removable anti-static comfort insole with moisture control system; orthopaedic insoles possible
  • Anti-slip sole with shock absorption at heel and front of the foot
  • Virtually seamless design to eliminate the potential for pressure points and blisters
  • Elastic laces for quick and individual adjustment
  • 100% metal-free xenova® toecap, non magnetic (200 J)
  • Fulfils ESD requirements: Resistance to ground inferior to 35 megaohms
Suitable for people allergic to chrome.
Colour: Black
Width: 10, 11, 12, 14
Certificeringer: EN 61340 (ESD); DGUV112-191; EN ISO 20345:2011; S2, SRC