Jars, wide mouth, with screw and inner cap
Leverandør: LAMAPLAST
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
22 23 53 24 26 25
215-4066EA 24.6 DKK
215-4066 215-4067 215-4070 215-4068 215-4069 215-4184
Jars, wide mouth, with screw and inner cap
Dåser Dåser, allround
HDPE, translucent, with black screw cap.
  • Good chemical and thermal resistance
  • Wide neck for easy filling and cleaning
  • With screw cap and inner cap
  • For food use
Equipped with inner cap and screw cap, they can be easily usable in various fields : from chemical, to animal husbrandy and agriculture, until the use as containers for cosmetics and pharmaceuticals, and many others. Thanks to the wide mouth, they’re suitable for the storage of : creams, semi-solid substances, crystals, granulates and powders.