Silica gels for preparative chromatography, low pressure, J.T.Baker®
Leverandør: Avantor
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
7024-00EA 3370 DKK
7024-00 7024-02 7024-05
Silica gels for preparative chromatography, low pressure, J.T.Baker®
Kromatografi sorbenter HPLC Sorbenter
These are used in low pressure preparative chromatography. The mesh specifications ensure there is no excess of fines in the product which can affect flow rates and separation performance.
  • Stort overfladeareal forbedrer produktivitet
  • Smal porestørrelsesfordeling forstærker spidser og forbedrer resolution