Dry box gloves, CSM
Leverandør: PIERCAN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
10800Y71/24/10 54800Y9.5E45 16800Y104/10 10800Y8.5E65 10800Y91/26/10 16800Y114/10 16800Y94/10
PIEC10800Y71/24/10EA 24640 DKK
PIEC10800Y71/24/10 PIEC54800Y9.5E45 PIEC16800Y104/10 PIEC10800Y8.5E65 PIEC10800Y91/26/10 PIEC16800Y114/10 PIEC16800Y94/10
Dry box gloves, CSM
Handsker
These gloves made of chlorosulphonated polyethylene provide excellent resistance to chemicals, sterilisation agents (hydrogen peroxide) and sterilisation processes (gamma and beta radiations).
  • High resistance against ozone and UV
  • Excellent flexibility and dexterity
  • Excellent resistance against ionising radiations
Ideal for life science applications (pharmaceutical/medical) and nuclear industries.
Colour: White
0,4 mm thickness: EN 388 (4XX1), EN 374 (AKL), EN 374-2003, EN 421; Cat. III
0,6 mm thickness: EN 388 (4X11), EN 374 (AKL), EN 374-2003, EN 421; Cat. III
Certificeringer: 0,4 mm thickness: EN 388:2016 (3XX1X), EN ISO 374-1 Type B (AMP), EN ISO 374-5 VIRUS, EN 421; Cat. III
0,6 mm thickness: EN 388:2016 (3X11X), EN ISO 374-1 Type B (AMP), EN ISO 374-5 VIRUS, EN 421; Cat. III