Hand cleansing wipes, pre-saturated, Sekuroka®
Leverandør: Roth Carl
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
CK45.1 CK46.1
ROTHCK45.1EA 0 DKK
ROTHCK45.1 ROTHCK46.1
Hand cleansing wipes, pre-saturated, Sekuroka®
Papirservietter Engangsservietter og - håndklæder Imprægnerede servietter
These pre-saturated PP tissues absorb and bind the dirt. They can be used repeatedly for the cleaning of hands, labware or tools.
  • Multi-purpose wipes: For a wide variety of applications; ideal for removing slight to medium soiling
  • Heavy-duty wipes: With additional rubbing effect for removing stubborn soiling, e.g. oil, fat and grease
  • Both qualities are lightly perfumed and feature a moisturising glycerine additive
  • Supplied in resealable bucket or canister
Advarsel: *Produktet/produkterne på denne side er uden for anvendelsesområdet for EU -direktiv 2019/904