ESD safety ankle boots, lace-up, Anatom, 32268
Leverandør: Abeba
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
32268/41 32268/36 32268/43 32268/47 32268/48 32268/49 32268/50 32268/51 32268/37 32268/38 32268/39 32268/40 32268/42 32268/44 32268/45 32268/46 32268/52
ABEB32268/41EA 837 DKK
ABEB32268/41 ABEB32268/36 ABEB32268/43 ABEB32268/47 ABEB32268/48 ABEB32268/49 ABEB32268/50 ABEB32268/51 ABEB32268/37 ABEB32268/38 ABEB32268/39 ABEB32268/40 ABEB32268/42 ABEB32268/44 ABEB32268/45 ABEB32268/46 ABEB32268/52
ESD safety ankle boots, lace-up, Anatom, 32268
Sko og støvler
ESD ankle boot made from smooth leather, reflective material and a fuel resistant PU outsole with high slip resistance and a self-cleaning profile.
  • Perspiration-absorbent special lining with silver fibre
  • Replaceable acc Wave insole; orthopaedic insoles possible
  • Sanitized® treatment reduces odour and bacterial growth
  • Asymmetric heel and Flexzone for a natural bedding and foot movement
  • TPU torsion waist, arch support and high stability
  • Shock-absorbent PU midsole
  • Metal-free puncture resistant midsole
  • Protective steel toecap (200 J)
Colour: White/grey
Certificeringer: EN 61340 (ESD); DGUV 112-191; EN ISO 20345:2011; S3, SRC