VWR® shiroGEL vertical 20, Vertical Electrophoresis Systems
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
700-0353EA 16190 DKK
700-0353 700-0354
VWR® shiroGEL vertical 20, Vertical Electrophoresis Systems
Elektroforesesystemer Vertikale Gelelektroforesesystemer
The shiroGEL vertical 20 offers application flexibility and is distinguished by its ease of gel casting and set-up, requiring only four screws to secure up to four 20×20 cm gels. This vertical screw clamp technology distributes pressure evenly along the height of gel rather than in the centre to eliminate plate bowing and gel compression, while maintaining a leakproof seal during casting. A built-in inner buffer chamber within the PAGE insert allows set up to be completed without inclusion of an upper buffer chamber. These systems are ideal for 1D and 2D SDS-PAGE, native, preparative,More Product Information
Bestil nu
Specifikationer
Environmentally Preferable
Gelstørrelse B×H 200×200 mm