Safety ankle boots, lace-up, Athletic Fresco
Leverandør: HONEYWELL SAFETY
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
6243855/37 6243855/38 6243855/39 6243855/40 6243855/41 6243855/42 6243855/43 6243855/44 6243855/45 6243855/46 6243855/47
DALL6243855/37EA 1140 DKK
DALL6243855/37 DALL6243855/38 DALL6243855/39 DALL6243855/40 DALL6243855/41 DALL6243855/42 DALL6243855/43 DALL6243855/44 DALL6243855/45 DALL6243855/46 DALL6243855/47
Safety ankle boots, lace-up, Athletic Fresco
Sko og støvler
Safety ankle boots made from suede and mesh upper, 3-D lining, watertight bellows plus PU/TPU sole.
  • Breathable, absorbent, fast drying Poromax® 3-D lining
  • Coolmax® insole for an optimal perspiration absorption
  • Shock absorbent PU/TPU sole
  • Metal-free, flexible, puncture resistant FLEXIUM ™ midsole with thermal insulation
  • Ultra-light 200 Joules Athletic™ metal-free toecap
  • Toe and heel reinforcements
Colour: Beige/grey
Certificeringer: EN ISO 20345:2004; S1P, CI, SRC, CI.