Media/storage bottles, square, graduated, with plug seal screw cap, PYREX®
Leverandør: Corning
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
431533 431532 431531 431530
CORN431533EA 2380 DKK
CORN431533 CORN431532 CORN431531 CORN431530
Media/storage bottles, square, graduated, with plug seal screw cap, PYREX®
Flasker Medieflasker
PET, clear.
  • Good for storage of media, buffers and other aqueous solutions
  • Large 37 mm opening provides easier access for liquid transfer
  • Screened white enamel graduations are easier to read than moulded graduations
  • Large white marking spot for easier identification and easy opening linear tear packaging
  • Individually packaged bottles are sterilised by gamma radiation and are nonpyrogenic
These storage bottles are easier to handle, require less space (13 to 20%) on the shelf and are ideal for mixing, sampling and storage. PET resin is lightweight,More Product Information