Heat resistant gloves, KarboTECT® L 954
Leverandør: KCL by Honeywell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
954.9 954.10
KCHL954.9EA 961 DKK
KCHL954.9 KCHL954.10
Heat resistant gloves, KarboTECT® L 954
Handsker
This ambidextrous glove is made of knitted para-aramide and contains carbon. This combination enables the glove wearer to work with temperatures of up to 350 °C.
  • Extended leather cuff ensures additional protection of the forearm
  • Five-finger-design offers a good flexibility and grip
  • Para-aramide can be washed several times without losing its mechanical protection properties
  • Good thermal insulation, high cut resistance
  • Reduction of electrostatic charge
  • High wearing comfort due to cotton inlay
Ideal for use in metal work and processing, large bakeries, refineries and other areas with dry heat contact. Can also be used when changing tools at hot machines or when working with heat ovens and cabinet dryers.
Colour: Brown/green
EN 388 (1441), EN 407 (4243XX); Cat. III
Certificeringer: EN 388:2016 (1X4XC), EN 407 (4242XX); Cat. III
Food contact approved