Safety ankle boots, Velcro® fastening, HACCP, Anatom, 2280
Leverandør: Abeba
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2280/39 2280/37 2280/40 2280/41 2280/42 2280/44 2280/45 2280/46 2280/47 2280/48
ABEB2280/39EA 850 DKK
ABEB2280/39 ABEB2280/37 ABEB2280/40 ABEB2280/41 ABEB2280/42 ABEB2280/44 ABEB2280/45 ABEB2280/46 ABEB2280/47 ABEB2280/48
Safety ankle boots, Velcro® fastening, HACCP, Anatom, 2280
Sko og støvler
Anti-static ankle boot made from microfibre, reflective material, Velcro® fastening, cushioned ankle support and a fuel resistant PU outsole with high slip resistance and a self-cleaning profile. Washable at 30 °C.
  • Cold insulating lining
  • Cushioned ankle support
  • Reflective stripes
  • Replaceable acc Wave insole; orthopaedic insoles possible
  • Sanitized® treatment reduces odour and bacterial growth
  • Asymmetric heel and Flexzone for a natural bedding and foot movement
  • TPU torsion waist, arch support and high stability
  • Shock-absorbent PU midsole
  • Metal-free puncture resistant midsole
  • Protective steel toecap (200 J)
Suitable for use in kitchens.
Colour: White
Certificeringer: DGUV 112-191; HACCP; EN ISO 20345:2011; S3, SRC.