Denatured ethanol, filtered, for cleanroom disinfection
Leverandør: CONTEC
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
FBC570DE FBT170DE
141-1371EA 1350 DKK
141-1371 141-1372
Denatured ethanol, filtered, for cleanroom disinfection
Renseopløsninger Disinfektanter
Ethanol 70% with 30% purified water, denatured with isopropyl alcohol.
  • The alcohol/water blend is 0,2 µm filtered
  • GMP manufactured under Grade A laminar flow in a Grade C cleanroom
Contec sterile alcohol is provided sterile by gamma irradiated using a validated process at no less than 25 kGy. Guaranteed endotoxin levels of \<0,25 EU/ml
Emballering: Double bagged packed in linear tear packaging.
Advarsel: Use biocides safely. Always read the label and product information before use.