Flasker, smal hals, med standard jordprop, BIBBY®
Leverandør: SciLabware
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SBA104 SBA100 SBA102 SBA106 SBC100 SBC102 SBC104
201-0782EA 909 DKK
201-0782 201-0780 201-0781 201-0783 201-0785 201-0786 201-0787
Flasker, smal hals, med standard jordprop, BIBBY®
Flasker Glasflasker
Soda-lime glass, clear or brown, with standard ground joint and polyethylene stopper.
  • Good chemical and physical resistance
  • For use in applications that are not associated with high thermal loads
  • Stable and robust
Available in crystal clear glass or brown tinted to protect light-sensitive substances.
Complies with ISO 4796-2
Certificeringer: Complies with ISO 4796-2