Capillary columns, CP-Al₂O₃/Na₂SO₄
Leverandør: VARIAN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
CP7565 CP7568 CP7567 CP7565I5 CP7586 CP7587
568-0707EA 21480 DKK
568-0707 568-0710 568-0709 568-0708 568-0711 568-0712
Capillary columns, CP-Al₂O₃/Na₂SO₄
Kromatografikolonner
CP-Al₂O₃/Na₂SO₄ is an aluminum oxide PLOT column that offers high selectivity for separating ppm levels of C₁ to C₅ hydrocarbons in a main stream of C₁ to C₅ hydrocarbons. The Na₂SO₄ deactivation salt provides a polar Al₂O₃ surface, so that unsaturated compounds such as ethylene and acetylene are retained longer. These columns analyse more compounds in a single run than packed columns, while still delivering higher resolution and faster analysis times. When compared to liquid stationary phases, CP-Al₂O₃ provides increased selectivity and allows all C₁ to C₅ hydrocarbon isomers to be separated. CP-Al₂O₃ operates without the need for sub-ambient cooling.
  • High capacity thick films
  • No need for sub-ambient cooling simplifies operation
Temperature limits: –100 to +200/200 °C
Similar phases: Al₂O₃/KCl, Rt-Alumina PLOT, Alumina PLOT, RT-Alumina BOND/KCl , Alumina chloride PLOT, AB-PLOT Al₂O₃ KCl