Gel permeation chromatography columns, PLgel MIXED
Leverandør: VARIAN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
PL1110-6100 PL1110-6500 PL1110-6200 PL1110-6300 PL1110-6504
VARIPL1110-6100EA 28020 DKK
VARIPL1110-6100 568-0784 568-0782 568-0783 568-0785
Gel permeation chromatography columns, PLgel MIXED
Kromatografikolonner
Using these mixed columns you can eliminate mismatched column sets and spurious peaks for more reliable results. Every column contains a mixture of individual pore size materials, accurately blended to cover a specified broad range of molecular weight with a linear calibration to eliminate column mismatch. Simply add extra columns for even greater resolution.