Gel permeation chromatography columns, PLgel individual pore size
Leverandør: VARIAN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
PL1110-6320 PL1110-1120 PL1110-1520 PL1110-1320 PL1110-6520 PL1110-6525 PL1510-1520 PL1510-5320 PL1110-6115 PL1110-6120 PL1110-6125 PL1110-6130 PL1110-6140 PL1110-6150 PL1110-6160 PL1110-6515 PL1110-6530 PL1110-6540 PL1110-6550 PL1110-8520
VARIPL1110-6320EA 36550 DKK
VARIPL1110-6320 568-0772 568-0774 568-0773 568-0787 568-0788 VARIPL1510-1520 VARIPL1510-5320 568-0775 568-0776 568-0777 568-0778 568-0779 568-0780 568-0781 568-0786 VARIPL1110-6530 VARIPL1110-6540 VARIPL1110-6550 VARIPL1110-8520
Gel permeation chromatography columns, PLgel individual pore size
Kromatografikolonner
Individual pore size GPC columns offer high resolution over a specific molecular weight range. The linear portion of the calibration curve, where the slope is at its shallowest, defines the MW region over which optimum resolution will be achieved.
  • Very high efficiency
  • Fast analysis