Safety shoes, Velcro® fastening, Protektor Line, 2230/2232
Leverandør: Abeba
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2232/47 2232/36 2232/37 2232/38
ABEB2232/47EA 558 DKK
ABEB2232/47 ABEB2232/36 ABEB2232/37 ABEB2232/38
Safety shoes, Velcro® fastening, Protektor Line, 2230/2232
Sko og støvler
Anti-static shoes made from smooth leather upper, leather lining, and a non slip PU sole.
  • PU sole with high slip resistance
  • Protective steel toecap (200 J)
  • Puncture resistant steel midsole
  • Double Velcro® fastening
Suitable for use in kitchens.
Colour: White or black
Certificeringer: EN ISO 20345:2011; S3, SRC