Safety shoes, lace-up, Athletic Fresco
Leverandør: HONEYWELL SAFETY
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
6243854/39 6243854/37 6243854/38 6243854/40 6243854/41 6243854/42 6243854/43 6243854/44 6243854/45 6243854/46 6243854/47
114-6938EA 1080 DKK
114-6938 114-6936 114-6937 114-6939 114-6940 114-6941 114-6942 114-6943 114-6944 114-6945 114-6946
Safety shoes, lace-up, Athletic Fresco
Sko og støvler
Safety shoes made from suede leather and mesh upper, 3-D lining plus a PU/TPU sole.
  • Breathable, absorbent, fast drying Poromax® 3-D lining
  • Shock-absorbent PU/TPU sole
  • Metal-free, flexible, puncture resistant FLEXIUM ™ midsole with thermal insulation
  • Ultra-light 200 Joules Athletic™ metal-free toecap
  • Forefoot and heel reinforcements
Colour: Black/grey.
Certificeringer: EN ISO 20345:2004; S1P, CI, SRC