High visibility traffic jacket, Lite Two-Tone, S166
Leverandør: Portwest
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
S166YNRS S166YNRM S166YNRL S166YNRXXXL S166YNRXL S166YNRXXL
PRTWS166YNRSEA 444 DKK
PRTWS166YNRS PRTWS166YNRM PRTWS166YNRL PRTWS166YNRXXXL PRTWS166YNRXL PRTWS166YNRXXL
High visibility traffic jacket, Lite Two-Tone, S166
Arbejdstøj
This two-tone jacket is made of 150D PVC coated Oxford weave polyester and features a polyester mesh lining.
  • Zipped front with Velcro storm flap
  • Jetted hip pockets
  • Stud adjustable cuffs
  • Concealed hood
  • Radio loop
  • Twin stitching on the critical seams
Colour: Hi-vis yellow/navy
Weight: 250 g/m² (body); 60 g/m² (Mesh lining)
Certificeringer: EN 343 (3-1), EN ISO 20471 (Class 3)