Industrial scales, Valor® 4000
Leverandør: OHAUS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
30035653 30035654 30035655 30035656 30035663 30035664 30035665 30035666 30072307 30072308 30072309 30072310 30072311 30072312 30072313 30072314
OHAU30035653EA 4810 DKK
OHAU30035653 OHAU30035654 OHAU30035655 OHAU30035656 611-3708 611-3709 611-3710 611-3711 611-4380 611-4381 611-4382 611-4383 611-4384 611-4385 611-4386 611-4387
Industrial scales, Valor® 4000
Vægte
Waterproof compact scales designed for the food industry. Features include keypad with four mechanical switches, adjustable feet, level bubble and cover with spring to protect power connector of the scale from ingress of moisture, especially when the scale is washed down. Dual LED display back and front enable two operators to work with the Valor® 4000 at the same time.
  • Stabilisation times of 0,5 s
  • Flow Thru™ design protects scales from water damage - water drains away ensuring the scale remains clean and operational
  • Touchless infrared sensor at the rear