High visibility fleece jackets, F300
Leverandør: Portwest
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
F300YERXS F300YERS F300YER5XL F300YERXXL F300YERXL F300YERM F300YERL F300YERXXXL F300YER4XL
PRTWF300YERXSEA 453 DKK
PRTWF300YERXS PRTWF300YERS PRTWF300YER5XL PRTWF300YERXXL PRTWF300YERXL PRTWF300YERM PRTWF300YERL PRTWF300YERXXXL PRTWF300YER4XL
High visibility fleece jackets, F300
Arbejdstøj
This jacket is made of 100% polyester with anti-pilling finish. It features a nylon mesh lining in the body and nylon lining in the sleeves. Ideal for cold and windy days.
  • With reflective tape for enhanced visibility
  • Anti-pilling finish ensures no unsightly bobbling on the surface of the fabric
  • Internal patch pocket, two secure side pockets with zips
  • Elasticated drawcord waist for more comfortable fit
Weight (outer fabric): 300 g/m²
Weight (polyester mesh - body): 60 g/m²
Weight (polyester taffeta - sleeves): 60 g/m²
Certificeringer: EN ISO 20471 (Class 3)