Leather gloves, Tergsus A250
Leverandør: Portwest
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
A250RERXL A250BLUM A250YERL A250BKRXXL
112-5139EA 61.2 DKK
112-5139 112-5137 112-5138 112-5140
Leather gloves, Tergsus A250
Handsker
Durable pig grain leather gloves with cotton back.
  • Velcro fastening at the cuff
  • Cotton back for improved breathability
  • Reinforced finger tips
Suitable for light industrial applications.
Certificeringer: EN 388 (3142X)