Pipette containers, VARICON
Leverandør: schuett-biotec GmbH
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
3521233 3521423
612-1850EA 510 DKK
612-1850 612-1851
Pipette containers, VARICON
Bokse Beholdere til Pipettesterilisering
These containers are made of two overlapping aluminium parts, which can be pushed in and out of each other in telescope fashion to match the length/voluminal of the pipettes.