Microcentrifuge tubes, with cap, SafeFit
Leverandør: BOETTGER PAUL
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
02.043.0100 02.053.0100 02.063.0100 02.053.0102 02.053.0104 02.043.0102 02.043.0103 02.043.0104
BOET02.043.0100EA 315 DKK
BOET02.043.0100 BOET02.053.0100 BOET02.063.0100 BOET02.053.0102 BOET02.053.0104 BOET02.043.0102 BOET02.043.0103 BOET02.043.0104
Microcentrifuge tubes, with cap, SafeFit
Rør Mikrocentrifugerør
Made of selected, especially clear polypropylene, optimal visibility of contents.
  • High Speed Protected: special stability for highest security in centrifugation
  • Special closing mechanism prevents accidental popping open of the cap
  • Tight closure such as for applications under thermal load (incubation)
  • Precise sealing for low evaporation rate