Wafer tweezers, Erem®
Leverandør: Basan
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
124542 600JSA
BSNC124542EA 617 DKK
BSNC124542 BSNC600JSA
Wafer tweezers, Erem®
Pincet
Stainless steel. These tweezers are suitable for 4 to 6" wafers. They feature a flat lower paddle and either 6 upper fingers or a free-floating Teflon® upper paddle.
  • Serrated finger grips for secure handling
  • Anti-magnetic, non rusting
  • Acid-proof, heat resistant surface
  • Model with 6 upper fingers for protecting wafers against damage
  • Model with free-floating Teflon upper paddle for secure, damage-free gripping